Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of APLF only when phosphorylated at Ser116.
C2ORF13 is a component of the cellular response to chromosomal DNA single- and double-strand breaks.[supplied by OMIM, Mar 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aprataxin and PNK-like factor; aprataxin- and PNK-like factor; Apurinic-apyrimidinic endonuclease APLF; PNK and APTX-like FHA domain-containing protein; PNK and APTX-like FHA protein; XRCC1-interacting protein 1; zinc finger, CX5CX6HX5H motif containing 1
Gene Aliases: 2010301N04Rik; AI452191; APFL; APLF; C2orf13; PALF; RGD1565557; XIP1; ZCCHH1
UniProt ID: (Human) Q8IW19, (Mouse) Q9D842
Entrez Gene ID: (Human) 200558, (Rat) 500247, (Mouse) 72103
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support