Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATRIP is an 86 kDa protein that is phosphorylated by ATR, regulates ATR expression, and is an essential component of the DNA damage checkpoint pathway. It localizes to the intranuclear foci after DNA damage or inhibition of replication.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATM and Rad3 related interacting protein; ATM and Rad3-related-interacting protein; ATR IP; ATR-interacting protein; DRN3; FLJ12343
Gene Aliases: 6620401K05Rik; AGS1; ATRIP
UniProt ID: (Human) Q8WXE1, (Mouse) Q8BMG1
Entrez Gene ID: (Human) 84126, (Rat) 301014, (Mouse) 235610
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support