Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of Afadin only when phosphorylated at Ser1799.
AFDN belongs to an adhesion system, probably together with the E-cadherin-catenin system, which plays a role in the organization of homotypic, interneuronal and heterotypic cell-cell adherens junctions (AJs). Nectin- and actin-filament-binding protein that connects nectin to the actin cytoskeleton.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Afadin; Afadin adherens junction formation factor; ALL1-fused gene from chromosome 6 protein; FLJ34371; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 4; Protein AF-6; RP3-431P23.3; RP3-470B24.4
Gene Aliases: AF6; AFDN; MLL-AF6; MLLT4
UniProt ID: (Human) P55196
Entrez Gene ID: (Human) 4301
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support