Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody detects endogenous protein at a molecular weight of 78 kDa.
Purity is >95% by SDS-PAGE.
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DCLRE1C protein; DNA cross-link repair 1A, PSO2 homolog; DNA cross-link repair 1C (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1C protein; FLJ11360; hSNM1C; mArt; Protein A-SCID; Protein artemis; RS SCID; severe combined immunodeficiency, type a (Athabascan); SNM1 homolog C; SNM1-like protein; SNM1C antibody; tein
Gene Aliases: 9930121L06Rik; A-SCID; AI661365; Art; ARTEMIS; ASCID; DCLRE1C; DCLREC1C; RS-SCID; SCIDA; SNM1C; Snm1l
UniProt ID: (Human) Q96SD1, (Mouse) Q8K4J0, (Rat) Q5XIX3
Entrez Gene ID: (Human) 64421, (Mouse) 227525, (Rat) 259171
Molecular Function: DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support