Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Predicted to react with Mouse and Rat samples.
Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cdc2-related protein kinase; CDK1; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; Cell division control protein 2 homolog; cell division cycle 2 homolog A; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; Cell division protein kinase 1; Cell division protein kinase 2; cyclin dependent kinase 2-alpha; Cyclin-dependent kinase 1; Cyclin-dependent kinase 2; p33 protein kinase; p34 protein kinase
Gene Aliases:
A630093N05Rik; CDC2; CDC28A; Cdc2a; CDK1; CDK2; CDKN1; CDKN2; p33(CDK2); p34
UniProt ID: (Human) P06493, (Human) P24941, (Rat) P39951, (Mouse) P11440, (Mouse) P97377
Entrez Gene ID: (Human) 983, (Human) 1017, (Rat) 54237, (Mouse) 12534, (Rat) 362817, (Mouse) 12566
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support