Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Calcineurin is a Ser/Thr, calcium and calmodulin-dependent protein phosphatase that plays an essential role in the T cell activation pathway. Calcineurin (also referred to as protein phosphatase 2B) is composed of two subunits; calcineurin A (CnA), a 60 kDa catalytic subunit which is highly homologous to protein phosphatases 1 and 2A; and a 19 kDa regulatory subunit, calcineurin B (CnB) which, like calmodulin, contains four EF-hand calcium binding sites. Calcineurin-mediated dephosphorylation of the nuclear factor of activated T-cells (NF-AT) is essential for NF-AT activation, nuclear translocation, and early gene expression in T-cells. The immunosuppressive drugs, cyclosporin A and FK506, when bound to the immunophilins, cyclophilin and FKBP respectively, inhibit dephosphorylation and activation of NF-AT by calcineurin. This inhibition is caused by the immunosuppressant/immunophilin complex simultaneously binding to both CnA and CnB which does not inhibit the phosphatase activity of CnA, but rather blocks access of the substrate to the catalytic domain of CnA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALNB1; calcineurin B, type I (19kDa); Calcineurin subunit B type 1; PP2B beta 1; protein phospatase 3, regulatory subunit B, alpha isoform (calcineurin B, type I); protein phospatase 3, regulatory subunit B, alpha isoform,type 1; Protein phosphatase 2B regulatory subunit 1; protein phosphatase 2B regulatory subunit B alpha; protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform (calcineurin B, type I); protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform (calcineurin B, type I); protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform; protein phosphatase 3 regulatory subunit B alpha; Protein phosphatase 3 regulatory subunit B alpha isoform 1; protein phosphatase 3, regulatory subunit B, alpha; protein phosphatase 3, regulatory subunit B, alpha isoform (calcineurin B, type I)
Gene Aliases: CALNB1; CaNB1; CNA2; CNB; CNB1; MCIP1; PPP3R1
UniProt ID: (Human) P63098, (Mouse) Q63810, (Rat) P63100
Entrez Gene ID: (Human) 5534, (Mouse) 19058, (Rat) 29748
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support