Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Antibody detects endogenous levels of Calmodulin1/2/3 only when phosphorylated at Thr80 + Ser82, which site historically referenced as Thr79+Ser81.
Calmodulin is a small, highly conserved calcium binding protein found in all eukaryotic cells. With the capacity to bind up to four calcium ions, this 17 kDa protein acts as an important intracellular receptor for regulatory calcium signals. As it binds calcium, calmodulin undergoes conformational changes which can increase its affinity for target proteins. It acts both directly, through interaction with key target enzymes, and indirectly, via specific kinases. Studies have found that calmodulin participates in the regulation of several biological processes including energy and biosynthetic metabolism, cell motility, exocytosis, cytoskeletal assembly, and intracellular modulation of both cAMP and calcium concentrations.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calmodulin; Calmodulin 1 (phosphorylase kinase, delta); calmodulin 2 (phosphorylase kinase, delta); calmodulin 3 (phosphorylase kinase, delta); Calmodulin III; Calmodulin-1; Calmodulin-2; Calmodulin-3; caM; epididymis secretory protein Li 72; LP7057 protein; phosphorylase kinase delta; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 2; prepro-calmodulin 3
Gene Aliases: 1500001E21Rik; AI256814; AI327027; AI461935; AL024000; AL024017; CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CaMA; CAMB; CAMC; CAMI; CAMII; CAMIII; CPVT4; DD132; HEL-S-72; LQT14; LQT15; PHKD; PHKD2; PHKD3; R75142; RNCAMIII
UniProt ID: (Human) P0DP25, (Human) Q61379, (Rat) P0DP31, (Mouse) P0DP28
Entrez Gene ID: (Human) 801, (Human) 805, (Human) 808, (Rat) 24242, (Mouse) 12313, (Rat) 50663, (Mouse) 12314, (Rat) 24244, (Mouse) 12315
Molecular Function:
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support