Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
FIGURE: 1 / 2
Casein kinase I (also designated CKI) and casein kinase II (also designated CKII) compose a family of serine/ threonine protein kinases which are present in all eukaryotes examined to date. CKI family members, which include CKI alpha, gamma, epsilon and delta, have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. CKII is usually expressed as a tetrameric complex consisting of either an alpha2 beta2 or an alpha alpha' beta2 structure. The alpha catalytic subunit is stimulated by the beta regulatory subunit, which undergoes autophosphorylation. CKII activity is high in the cytosol and nucleus of proliferating and differentiating cells. CKII is known to phosphorylate more than 100 different substrates including nuclear oncoproteins, transcription factors and enzymes involved in DNA metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: casein kinase 1 gamma 2 isoform; casein kinase 1 isoform gamma-2; casein kinase 1, gamma 1; casein kinase 1, gamma 2; casein kinase 1, gamma 3; Casein kinase I isoform gamma-1; Casein kinase I isoform gamma-2; Casein kinase I isoform gamma-3; casein kinase I, gamma 1; CK I gamma1; CK-I gamma1; CK1G1; CKI gamma 1; CKI gamma-1; CKI gamma1; CKI- gamma1; CKI-gamma 1; CKI-gamma 2; CKI-gamma 3; CKI-gamma-1; KC1G1
Gene Aliases: 2810429I12Rik; 3300002K07Rik; 9130020E21Rik; AI463719; C330049O21Rik; CK1G2; CK1gamma1; CKI-gamma 3; CSNK1G1; CSNK1G2; CSNK1G3; CSNK1G3L
UniProt ID: (Human) Q9HCP0, (Human) P78368, (Human) Q9Y6M4, (Mouse) Q8BTH8, (Rat) Q62761, (Mouse) Q8BVP5, (Rat) Q62762, (Mouse) Q8C4X2, (Rat) Q62763
Entrez Gene ID: (Human) 53944, (Human) 1455, (Human) 1456, (Mouse) 214897, (Rat) 64086, (Mouse) 103236, (Rat) 65278, (Mouse) 70425, (Rat) 64823
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support