Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The antibody detects a 60 kDa* band corresponding to Coronin-1B on SDS-PAGE immunoblots of human A431 or HeLa cells treated with Calyculin A. This band is not observed in control cells and the band observed in Calyculin A-treated cells can be removed with lambda phosphatase treatment.
Members of the coronin family, such as CORO1B, are WD repeat-containing actin-binding proteins that regulate cell motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coronin 1b; coronin 2; coronin, actin binding protein, 1B; Coronin-1B; Coronin-2
Gene Aliases: CORO1B; CORONIN-2
UniProt ID: (Human) Q9BR76, (Rat) O89046, (Mouse) Q9WUM3
Entrez Gene ID: (Human) 57175, (Rat) 29474, (Mouse) 23789
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support