Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Antibody detects endogenous levels of CUL4A only when phosphorylated at Ser40.
CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CUL 4A; CUL-4A; Cul4a protein; Cullin-4A; MGC36573; MGC64071; OTTHUMP00000018753
Gene Aliases: 2810470J21Rik; AW495282; CUL4A; RGD1563853
UniProt ID: (Human) Q13619, (Mouse) Q3TCH7
Entrez Gene ID: (Human) 8451, (Mouse) 99375, (Rat) 361181
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support