Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The laminar organization of multiple neuronal types in the cerebral cortex is required for normal cognitive function. In mice, the disabled-1 gene plays a central role in brain development, directing the migration of cortical neurons past previously formed neurons to reach their proper layer. This gene is similar to disabled-1, and the protein encoded by this gene is thought to be a signal transducer that interacts with protein kinase pathways to regulate neuronal positioning in the developing brain. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dab reelin signal transducer 1; Dab, reelin signal transducer, homolog 1; Disabled homolog 1; RP23-356E21.1; yotari
Gene Aliases: AI956902; C630028C02Rik; DAB1; scm; scr; scrambler; yot
UniProt ID: (Human) O75553, (Rat) Q8CJH2, (Mouse) P97318
Entrez Gene ID: (Human) 1600, (Rat) 266729, (Mouse) 13131
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support