Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 12
It is not recommended to aliquot this antibody.
This antibody is not cross-reactive with the corresponding phosphorylated residues of either JNK/SAPK or p38 MAP kinase.
ERK1 and ERK2 are widely expressed and are involved in the regulation of meiosis, mitosis, and postmitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. When growth factors bind to the receptor tyrosine kinase, Ras interacts with Raf, the serine/threonine protein kinase and activates it as well. Once actived, Raf phosphorylates serine residue in 2 further kinases, MEK1/2, which in turn phosphorylates tyrosine/threonine in extracellular-signal regulated kinase (ERK) 1/2. Upon activation, the ERKs either phosphorylate a number of cytoplasmic targets or migrate to the nucleus, where they phosphorylate and activate a number of transcription factors such as c-Fos and Elk-1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; Diphospho-ERK; dp-ERK; dpERK; drosophila ERK; enhancer of seven in absentia 7; ERK; Erk MAP kinase; ERK-1; ERK-2; ERK/rolled; erk1 erk2; ERK1b; ERK2; ERT1; ERT2; extracellular signal-regulated kinase; Extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; Insulin-stimulated MAP2 kinase; MAP kinase; MAP kinase 1; MAP kinase 2; MAP kinase 3; MAP kinase isoform p42; MAP kinase isoform p44; MAP-kinase; MAPK; MAPK 1; MAPK 2; MAPK 3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase; mitogen activated protein kinase 1; mitogen-activated 3; mitogen-activated protein kinase; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; Mitogen-activated protein kinase 3; Mitogen-activated protein kinase ERK-A; MNK1; p42; p42 MAP Kinase; p42-MAPK; p44 MAP kinase; p44-ERK1; p44-MAPK; phospho-ERK; pp42/MAP kinase; protein kinase; Protein rolled; protein tyrosine kinase ERK2; RE08694p; rl-PA; rl-PC; rl-PD; rl-PE; rl-PG; rl-PH; rl-PI; roll; sevenmaker
Gene Aliases: 12559; 9030612K14Rik; AA407128; AU018647; BcDNA:RE08694; C78273; CG12559; CG18732; CT34260; CT39192; D-ERK; DERK; DERK-A; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmERK; DmERK-A; DmERKA; DmMAPK; dp-ERK; DpERK; dpERK1; dpMAPK; Dsor2; E(sina)7; EC2-1; EK2-1; ERK; ERK-1; ERK-2; ERK-A; Erk/Map kinase; ERK1; ERK1/2; ERK2; ERKA; ERT1; ERT2; Esrk1; EY2-2; fi06b09; GroupII; HS44KDAP; HUMKER1A; l(2)41Ac; l(2R)EMS45-39; MAP-k; MAPK; MAPK1; MAPK2; MAPK3; MNK1; Mtap2k; p-ERK; p38; p40; p41; p41mapk; p42-MAPK; P42MAPK; p44; p44-ERK1; p44-MAPK; P44ERK1; P44MAPK; pERK; pMAPK; PRKM1; PRKM2; PRKM3; RL; rl/ERK; rl/MAPK; rll; SEM; SR2-1; Su(Raf)2B; wu:fi06b09; zERK1; zERK2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support