Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
A suggested positive control for Western blot is HepG2 cells; suggested positive control for ICC/IF is Hela cells.
The Eph subfamily represents the largest group of receptor protein tyrosine kinases identified to date. While the biological activities of these receptors have yet to be determined, there is increasing evidence that they are involved in central nervous system function and in development. The Eph subfamily receptors of human origin (and their murine/avian homologs) include EphA1(Eph), EphA2 (Eck), EphA3 (Hek4), EphA4 (Hek8), EphA5 (Hek7), EphA6 (Hek12),EphA7 (Hek11/MDK1), EphA8 (Hek3), EphB1 (Hek6), EphB2 (Hek5), EphB3(Cek10, Hek2), EphB4 (Htk), EphB5 (Hek9) and EphB6 (Mep). Ligands for Eph receptors include ephrin-A4 (LERK-4) which binds EphA3 and EphB1. In addition, Ephrin-A2 (Elf-1) has been described as the ligand for EphA4, ephrin-A3 (Ehk1-L) as the ligand for EphA5 and ephrin-B2 (Htk-L) as the ligand for EphB4 (Htk).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EK4; EK8; EPH-like kinase 4; EPH-like kinase 8; Eph-like tyrosine kinase 1; Ephrin A2; Ephrin A3; Ephrin A4; Ephrin type-A receptor 2; Ephrin type-A receptor 3; Ephrin type-A receptor 4; EphrinA2; EphrinA3; EphrinA4; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; HEK; Human embryo kinase; human embryo kinase 1; receptor protein-tyrosine kinase HEK8; soluble EPHA2 variant 1; testicular tissue protein Li 64; TYRO1 protein tyrosine kinase; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor MPK-5; Tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase receptor SEK-2; Tyrosine-protein kinase TYRO1; Tyrosine-protein kinase TYRO4
Gene Aliases: 2900005C20Rik; AI385584; ARCC2; AW492086; AW545284; Cek4; Cek8; CTPA; CTPP1; CTRCT6; ECK; EK4; End3; EPHA2; EPHA3; EPHA4; ETK; ETK1; HEK; HEK4; HEK8; Mek4; Myk2; rb; SEK; Sek-2; Sek1; Sek2; TYRO1; TYRO4
UniProt ID: (Human) B5A968, (Human) P29320, (Human) P54764, (Mouse) Q03145, (Mouse) Q03137
Entrez Gene ID: (Human) 1969, (Human) 2042, (Human) 2043, (Mouse) 13836, (Mouse) 13837, (Mouse) 13838
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support