Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with bovine, human, mouse, Xenopus and zebrafish based on 100% sequence homology.
This antibody is specific for the ~46 Dak EphrinB protein phosphorylated at Tyr317. Immunolabeling can be blocked by lambda-phosphatase treatment.
EPHB2 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cEK5; chicken embryo kinase 5; EFL6; EFNB3; EK5; embryo kinase 5 protein CEK5; EPH B2; EPH-like kinase 5; EPHB3; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin B4; Ephrin type-B receptor 2; EphrinB1; EphrinB2; EphrinB3; EphrinB4; EPLG8; ETK2; HEK-6; HEK2; LERK8; MGC87492; Neural kinase; Nuk receptor tyrosine kinase; TYRO6; Tyrosine-protein kinase receptor EPH-3; Tyrosine-protein kinase receptor SEK-3
Gene Aliases: CEK5; Drt; EPHB2; Epth3; Erk; ETECK; Hek5; Nuk; Prkm5; Qek5; RGD1564232; Sek3; Tyro5
UniProt ID: (Chicken) P28693, (Mouse) P54763
Entrez Gene ID: (Chicken) 396513, (Mouse) 13844, (Rat) 313633
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support