Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: AAYTT; Positive Samples: 293T, Hela; Cellular Location: Cell membrane, Single-pass type I membrane protein
EPHA3 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA3 encodes a protein that binds ephrin-A ligands and receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EPHA3 binds ephrin-A ligands.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EK4; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; H TYRO4; HEK; Human embryo kinase; human embryo kinase 1; testicular tissue protein Li 64; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase TYRO4
Gene Aliases: EK4; EPHA3; ETK; ETK1; HEK; HEK4; TYRO4
UniProt ID: (Human) P29320
Entrez Gene ID: (Human) 2042
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support