Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 11
Positive Control: HeLa starved for 3 hours then add 100 nM Calyculin A for 30 minutes cell lysate, NIH/3T3 starved for 24 hours then add 100 nM Calyculin A for 30 minutes cell lysate, C6 treated with 100 ng/mL Calyculin A for 1 hours cell lysate, mouse liver tissue, rat liver tissue.
Tissue Specificity: Low tissue specificity.
Subcellular Location: Cell membrane. Cell projection. Cytoplasm. Cytoskeleton. Membrane.
Sequence Similarities: 100% Mouse/Rat.
Predicted band size: 69 kDa.
Moesin, a member of the talin-4.1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytovillin; cytovillin 2; epididymis luminal protein 70; epididymis secretory protein Li 105; ESP10; Ezrin; Membrane-organizing extension spike protein; Moesin; p81; Radixin; radixin isoform; villin 2; villin 2 (ezrin); Villin-2
Gene Aliases: AA516625; AW146364; C78546; CVIL; CVL; DFNB24; EZR; HEL-S-105; HEL70; MSN; p81; R75297; RDX; VIL2
UniProt ID: (Human) Q4VX75, (Mouse) P26040, (Rat) P31977, (Human) P26038, (Rat) O35763, (Mouse) P26041, (Human) P35241, (Mouse) P26043
Entrez Gene ID: (Human) 7430, (Mouse) 22350, (Rat) 54319, (Human) 4478, (Rat) 81521, (Mouse) 17698, (Human) 5962, (Rat) 315655, (Mouse) 19684
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support