Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with bovine, human, mouse and non-human primate based on 100% sequence homology.
This antibody detects a band at ~102 kDa, representing GABAb R1 protein phosphorylated at Ser923.
Receptor for GABA. The activity of this receptor is mediated by G-proteins that inhibit adenylyl cyclase activity, stimulates phospholipase A2, activates potassium channels, inactivates voltage-dependent calcium-channels and modulates inositol phospholipids hydrolysis. Plays a critical role in the fine-tuning of inhibitory synaptic transmission. Pre-synaptic GABA-B-R inhibit neurotransmitter release by down-regulating high-voltage activated calcium channels, whereas postsynaptic GABA-B-R decrease neuronal excitability by activating a prominent inwardly rectifying potassium (Kir) conductance that underlies the late inhibitory postsynaptic potentials. Not only implicated in synaptic inhibition but also in hippocampal long-term potentiation, slow wave sleep, muscle relaxation and antinociception.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bM573K1.1.1 (gamma-aminobutyric acid (GABA) B receptor, 1a); DAAP-188P13.3; dJ271M21.1.1; dJ271M21.1.2; FLJ92613; GABA(B) receptor; GABA-B receptor 1; GABA-B-R1; Gaba-b-r1a; Gaba-b1; GABA-BR1; Gaba-br1a receptor; GABA-BR1b receptor; GABAB; GABABR1; Gababr1 receptor; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA-B) receptor, 1; Gamma-aminobutyric acid type B receptor subunit 1; gb1; GBR1; HGB1a; OTTHUMP00000109099; OTTHUMP00000214725
Gene Aliases: bM573K1.1; GABAB1; GABAbR1; Gabbr1
UniProt ID: (Mouse) Q9WV18, (Rat) Q9Z0U4
Entrez Gene ID: (Mouse) 54393, (Rat) 81657
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support