Product References
Vitamin D and rosuvastatin alleviate type-II diabetes-induced cognitive dysfunction by modulating neuroinflammation and canonical/noncanonical Wnt/β-catenin signaling.
PloS one
Muneeb M,Mansou SM,Saleh S,Mohammed RA
MA5-14873 was used in Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot to accentuated the neuroprotective potential of VitD and RSV and provide new incentives to expand their use in T2DM-induced cognitive and memory decline.
Wed Nov 16 00:00:00 EST 2022
The neuroprotective mechanism of lithium after ischaemic stroke.
Communications biology
Chen B,Zhang M,Ji M,Zhang D,Chen B,Gong W,Li X,Zhou Y,Dong C,Wen G,Zhan X,Wu X,Yuan H,Xu E,Xia M,Verkhratsky A,Li B
Published figure using Phospho-GSK3B (Ser9) monoclonal antibody (Product # MA5-14873) in Western Blot
Thu Feb 03 00:00:00 EST 2022
The neuroprotective mechanism of lithium after ischaemic stroke.
Communications biology
Chen B,Zhang M,Ji M,Zhang D,Chen B,Gong W,Li X,Zhou Y,Dong C,Wen G,Zhan X,Wu X,Yuan H,Xu E,Xia M,Verkhratsky A,Li B
Published figure using Phospho-GSK3B (Ser9) monoclonal antibody (Product # MA5-14873) in Western Blot
Thu Feb 03 00:00:00 EST 2022
The novel anti-colitic effect of β-adrenergic receptors via modulation of PS1/BACE-1/Aβ axis and NOTCH signaling in an ulcerative colitis model.
Frontiers in pharmacology
Nasser S,Abdallah DM,Ahmed KA,Abdel-Mottaleb Y,El-Abhar HS
MA5-14873 was used in Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot to investigate the impact of mirabegron (β3-agonist) and/or carvedilol (β1/β2 antagonist) on iodoacetamide-induced ulcerative colitis with emphasis on the novel pathomechanism of the PS1/BACE-1/Aβ axis in ulcerative colitis, and its relation to the inflammatory cascade, fibrotic processes, and the gut barrier dysfunction.
Fri Nov 18 00:00:00 EST 2022
MYBL2 in synergy with CDC20 promotes the proliferation and inhibits apoptosis of gastric cancer cells.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University
Deng Q,Wu L,Li Y,Zou L
MA5-14873 was used in Western Blot to examine the role of myeloblastosis viral oncogene homolog-like 2 (MYBL2) in gastric cancer progression and investigate the underlying mechanisms.
Wed Sep 01 00:00:00 EDT 2021
The extracellular HDAC6 ZnF UBP domain modulates the actin network and post-translational modifications of Tau.
Cell communication and signaling : CCS
Balmik AA,Sonawane SK,Chinnathambi S
Published figure using Phospho-GSK3B (Ser9) monoclonal antibody (Product # MA5-14873) in Immunocytochemistry
Sat May 01 00:00:00 EDT 2021
Sevoflurane inhibits the proliferation and invasion of hepatocellular carcinoma cells through regulating the PTEN/Akt/GSK‑3β/β‑catenin signaling pathway by downregulating miR‑25‑3p.
International journal of molecular medicine
Cao Y,Lv W,Ding W,Li J
MA5-14873 was used in Western Blotting to determine the molecular mechanisms governing sevoflurane's action in hepatocellular carcinoma cells.
Wed Jul 01 00:00:00 EDT 2020
ATM-associated signalling triggers the unfolded protein response and cell death in response to stress.
Communications biology
Hotokezaka Y,Katayama I,Nakamura T
MA5-14873 was used in Immunocytochemistry-immunoflourescence to study the molecular mechanisms involved in stress-induced apoptosis.
Tue Jul 14 00:00:00 EDT 2020
ATM-associated signalling triggers the unfolded protein response and cell death in response to stress.
Communications biology
Hotokezaka Y,Katayama I,Nakamura T
MA5-14873 was used in Immunocytochemistry-immunoflourescence to study the molecular mechanisms involved in stress-induced apoptosis.
Tue Jul 14 00:00:00 EDT 2020
Lithium treatment decreases activities of tau kinases in a murine model of senescence.
Journal of neuropathology and experimental neurology
Tajes M,Gutierrez-Cuesta J,Folch J,Ferrer I,Caballero B,Smith MA,Casadesus G,Camins A,Pallás M
MA5-14873 was used in western blot to study the effect of lithium on the acctivity of tau kinases in a mouse model of senescence
Sun Jun 01 00:00:00 EDT 2008