Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of Hic-5 only when phosphorylated at Tyr60.
This gene encodes a coactivator of the androgen receptor, a transcription factor which is activated by androgen and has a key role in male sexual differentiation. The encoded protein is thought to regulate androgen receptor activity and may have a role to play in the treatment of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Androgen receptor coactivator 55 kDa protein; androgen receptor coactivator ARA55; Androgen receptor-associated protein of 55 kDa; Hic-5; Hydrogen peroxide-inducible clone 5 protein; hydrogen peroxide-inducible clone-5; TGF beta-stimulated clone 5; Transforming growth factor beta-1-induced transcript 1 protein; TSC-5
Gene Aliases: ARA55; HIC-5; HIC5; TGFB1I1; TSC-5
UniProt ID: (Human) O43294, (Mouse) Q62219, (Rat) Q99PD6
Entrez Gene ID: (Human) 7041, (Mouse) 21804, (Rat) 84574
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support