Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This affinity-purified antibody is directed against phosphorylated human HMGN protein at pS20 and pS24 residues (other HMGN proteins have this conserved sequence but at other residue positions). Reactivity occurs against human HMGN pS20/pS24 proteins and the antibody is specific for the phosphorylated form of the proteins. Reactivity is seen against HMGN1 and HMGN2. HMGN3 is also likely reactive although not tested. Reactivity with non-phosphorylated human HMGNs is minimal by ELISA. A BLAST analysis was used to suggest cross reactivity with HMGN proteins from Xenopus, chicken, mouse, bovine, dog, orangutan, rat and swine based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
The high-mobility group (HMG) proteins 14 and 17 are abundant chromosomal proteins that bind to nucleosomes and enhance transcription. HMG-14 and HMG-17 also function as architectural elements, which alter the structure of the chromatin fiber and enhance transcription from chromatin templates. HMG-14/17 proteins modify the nucleosomal organization of the 30 nm chromatin fiber and mediate the unfolding of the higher order chromatin structure thereby facilitating access to the underlying DNA sequence. Clustering of architectural elements, such as HMG proteins and linker histone subtypes into distinct domains, may lead to structural and functional heterogeneity along the chromatin fiber. In addition, HMG-14 and HMG-17 have been identified as constitutive components of mouse oocyte and embryonic chromatin that establish a link between the structure of embryonic chromatin and the normal progression of embryonic development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ27265; FLJ31471; High mobility group nucleosome-binding domain-containing protein 1; high-mobility group (nonhistone chromosomal) protein 14; high-mobility group nucleosome binding 1; high-mobility group nucleosome binding domain 1; HMG 14; HMGN 1; MGC104230; MGC117425; Non-histone chromosomal protein HMG-14; nonhistone chromosomal protein HMG-14
Gene Aliases: HMG14; HMGN1
UniProt ID: (Human) P05114
Entrez Gene ID: (Human) 3150
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support