Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control for Western blot is HepG2 cells.
INSR is a receptor tyrosine kinase. Binding of insulin to the extracellular domain of INSR stimulates glucose uptake. INSR and IGF-1 receptors share major structural and functional similarity. The earliest cellular response to insulin stimulation is autophosphorylation of tyrosine in INSR. In humans, the INSR gene is located on chromosome 19. Defects in INSR are the cause of various insulin resistance syndromes and IGF-1R defects may also cause some forms of growth retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD 220; CD220; CD220 antigen; CD220 beta; HHF 5; Insulin receptor; IR; IR 1; IR beta; IR-1; IR1
Gene Aliases: CD220; HHF5; INSR
UniProt ID: (Human) P06213
Entrez Gene ID: (Human) 3643
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support