Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Immunogen sequence: NEYTA
LCK is a member of the Src-family tyrosine kinase, which are essential for signaling through the T-cell receptor (TCR) complex. Upon TCR triggering, LCK phosphorylates the ITAM motives in its zeta subunits establishing binding sites for the SH2 domains of the tyrosine kinase ZAP70, which is also phosphorylated by LCK and activated to generate subsequent signaling platforms by phosphorylation of adaptor LAT. The majority of LCK is localized to the plasma membrane, however, there is also a significant fraction associated with the Golgi apparatus which may contribute to Raf activation under conditions of weak stimulation through the TCR. LCK contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. LCK is also involved in the regulation of apoptosis induced by various stimuli, but not by the death receptors. Diseases associated with LCK include Immunodeficiency 22 and CD45 deficiency. Alternatively splice variants of the LCK gene encoding different isoforms of the protein have been found. Fyn is a membrane-associated non-receptor protein tyrosine kinase that belongs to the Src-family of kinases. The unique N-terminal domain of Fyn interacts with the CD3 and Delta chains of the TcR. Fyn is highly expressed in brain suggesting that it may have a role in the sensory nervous network and in myelination at early stages of CNS formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-syn protooncogene; FYN oncogene related to SRC, FGR, YES; Leukocyte C-terminal Src kinase; LSK; Lymphocyte cell-specific protein-tyrosine kinase; OKT3-induced calcium influx regulator; p56(LSTRA) protein-tyrosine kinase; p56-LCK; p59-Fyn; Protein YT16; Proto-oncogene c-Fyn; Proto-oncogene Lck; Proto-oncogene Syn; proto-oncogene tyrosine-protein kinase LCK; SLK; Src-like kinase; src/yes-related novel; T cell-specific protein-tyrosine kinase; T-lymphocyte specific protein tyrosine kinase p56lck; tyrosine kinase p59fyn(T); Tyrosine-protein kinase Fyn; Tyrosine-protein kinase Lck
Gene Aliases: FYN; IMD22; LCK; LSK; p56lck; p59-FYN; pp58lck; SLK; SYN; YT16
UniProt ID: (Human) P06241, (Human) P06239
Entrez Gene ID: (Human) 2534, (Human) 3932
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support