Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 18
Antibody detects endogenous levels of LIMK2 only when phosphorylated at Serine 283.
LIMK2 is a serine/threonine kinase involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin. LIMK2 has also been implicated in brain development. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. The protein encoded by LIMK2 is phosphorylated and activated by ROCK, a downstream effector of Rho, and the encoded protein, in turn, phosphorylates cofilin, inhibiting its actin-depolymerizing activity. It is thought that this pathway contributes to Rho-induced reorganization of the actin cytoskeleton. At least three transcript variants encoding different isoforms have been found for LIMK2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.11.1; kinase LIMK2; LIM domain kinase 2; LIM kinase 2; LIM motif-containing protein kinase 2; LIMK; LIMK 2; LIMK-2; LIMK1; LINhp-2; OTTHUMP00000199483
Gene Aliases: Limk-2; LIMK2; Limk2b; Link2
UniProt ID: (Human) P53671, (Mouse) O54785, (Rat) P53670
Entrez Gene ID: (Human) 3985, (Mouse) 16886, (Rat) 29524
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support