Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a member of the basic helix-loop-helix leucine zipper (bHLHZ) family of transcription factors. It is able to form homodimers and heterodimers with other family members, which include Mad, Mxi1 and Myc. Myc is an oncoprotein implicated in cell proliferation, differentiation and apoptosis. The homodimers and heterodimers compete for a common DNA target site (the E box) and rearrangement among these dimer forms provides a complex system of transcriptional regulation. Multiple alternatively spliced transcript variants have been described for this gene but the full-length nature for some of them is unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHd4; Class D basic helix-loop-helix protein 4; Delta-Max; MGC10775; MGC11225; MGC18164; MGC34679; MGC36767; Myc-associated factor X; Myc-binding novel HLH/LZ protein; orf1; Protein max; Protein myn
Gene Aliases: AA960152; AI875693; BHLHD4; bHLHd5; bHLHd6; bHLHd7; bHLHd8; MAX; Myn
UniProt ID: (Human) P61244, (Mouse) P28574, (Rat) P52164
Entrez Gene ID: (Human) 4149, (Mouse) 17187, (Rat) 60661
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support