Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 8
Antibody detects endogenous levels of MAP3K2 only when phosphorylated at Ser520.
MAP3K2 (MEKK2) is a serine/threonine kinase that activates kinases involved in the MAP kinase signaling pathway. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2.7.11.25; MAP/ERK kinase kinase 2; MAPK/ERK kinase kinase 2; MEK kinase 2; MEKK 2; mitogen activated protein kinase kinase kinase 2; Mitogen-activated protein kinase kinase kinase 2; protein kinase MEKK2b
Gene Aliases: 9630061B06Rik; AI585793; MAP3K2; MAPKKK2; MEKK2; MEKK2B
UniProt ID: (Human) Q9Y2U5
Entrez Gene ID: (Human) 10746, (Rat) 171492, (Mouse) 26405
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support