Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases, act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dual specificity mitogen-activated protein kinase kinase 3; Dual specificity mitogen-activated protein kinase kinase 6; MAP kinase kinase 3; MAP kinase kinase 6; MAPK/ERK kinase 3; MAPK/ERK kinase 6; MAPKK 3; MAPKK 6; MEK 3; MEK 6; mitogen activated protein kinase kinase 3; mitogen activated protein kinase kinase 6; OTTHUMP00000166044; protein kinase, mitogen activated, kinase 6; protein kinase, mitogen-activated, kinase 3; protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6); SAPK kinase 2; SAPK kinase 3; SAPKK3; Stress-activated protein kinase kinase 2; Stress-activated protein kinase kinase 3
Gene Aliases: AW212142; MAP2K3; MAP2K6; MAPKK3; MAPKK6; MEK3; MEK6; MKK3; MKK6; mMKK3b; PRKMK3; PRKMK6; SAPKK-2; SAPKK-3; SAPKK2; SAPKK3; SKK2; SKK3
UniProt ID: (Human) P46734, (Human) P52564, (Mouse) Q91VX1, (Mouse) P70236
Entrez Gene ID: (Human) 5606, (Human) 5608, (Mouse) 26397, (Rat) 303200, (Mouse) 26399, (Rat) 114495
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support