Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Antibody detects endogenous levels of MYPT1/2 only when phosphorylated at Ser668/Ser618.
Myosin phosphatase target subunit 1, which is also called the myosin-binding subunit of myosin phosphatase, is one of the subunits of myosin phosphatase. Myosin phosphatase regulates the interaction of actin and myosin downstream of the guanosine triphosphatase Rho. The small guanosine triphosphatase Rho is implicated in myosin light chain (MLC) phosphorylation, which results in contraction of smooth muscle and interaction of actin and myosin in nonmuscle cells. The guanosine triphosphate (GTP)-bound, active form of RhoA (GTP.RhoA) specifically interacted with the myosin-binding subunit (MBS) of myosin phosphatase, which regulates the extent of phosphorylation of MLC. Rho-associated kinase (Rho-kinase), which is activated by GTP. RhoA, phosphorylated MBS and consequently inactivated myosin phosphatase. Overexpression of RhoA or activated RhoA in NIH 3T3 cells increased phosphorylation of MBS and MLC. Thus, Rho appears to inhibit myosin phosphatase through the action of Rho-kinase. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hHS-M21A; MBSP; myosin binding subunit; Myosin phosphatase target subunit 1; Myosin phosphatase target subunit 2; myosin phosphatase, target subunit 1; myosin phosphatase, target subunit 2; Myosin phosphatase-targeting subunit 1; Myosin phosphatase-targeting subunit 2; PP-1M; Protein phosphatase 1 regulatory subunit 12A; Protein phosphatase 1 regulatory subunit 12B; protein phosphatase 1, regulatory (inhibitor) subunit 12A; protein phosphatase 1, regulatory (inhibitor) subunit 12B; protein phosphatase 1, regulatory subunit 12A; protein phosphatase 1, regulatory subunit 12B; Protein phosphatase myosin-binding subunit; Protein phosphatase subunit 1M; Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; smooth muscle myosin phosphatase myosin-binding subunit
Gene Aliases: 1200015F06Rik; 1810037O03Rik; 5730577I22Rik; 9530009M10Rik; AA792106; AI132431; AV099298; D10Ertd625e; M110; M130; MBS; MBSP; MYPT1; MYPT2; PP1bp55; PPP1R12A; PPP1R12B
UniProt ID: (Human) O14974, (Human) O60237, (Rat) Q10728, (Mouse) Q9DBR7
Entrez Gene ID: (Human) 4659, (Human) 4660, (Rat) 116670, (Mouse) 17931, (Mouse) 329251, (Rat) 304813
Molecular Function: phosphatase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support