Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
7TM Antibodies
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
MCHR1 is a member of the G protein-coupled receptor family 1. It is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 24; G-protein coupled receptor 24; GPCR24; MCH 1 receptor; MCH receptor 1; MCH-1R; MCH1; MCH1R; Melanin-concentrating hormone receptor 1; SLC-1; Somatostatin receptor-like protein
Gene Aliases: GPR24; MCH-1R; MCH1R; MCHR1; SLC-1; SLC1
UniProt ID: (Human) Q99705
Entrez Gene ID: (Human) 2847
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support