Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This gene encodes an F-box-containing protein that is a component of an SCF-type E3 ubiquitin ligase complex that regulates the onset of cell division. The G2/M transition in the cell cycle requires the interaction of the proteins cyclin B1 and cyclin-dependent kinase 1. The activated ubiquitin ligase complex targets the protein cyclin B1 for degradation, preventing this transition to mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3HC type 1; hematopoietic stem/progenitor cell protein 216; hNIPA; mNIPA; nuclear interacting partner of ALK; nuclear interacting partner of anaplastic lymphoma kinase (ALK); Nuclear-interacting partner of ALK; Nuclear-interacting partner of anaplastic lymphoma kinase; OTTHUMP00000212330; Zinc finger C3HC-type protein 1; zinc finger, C3HC-type 1; zinc finger, C3HC-type containing 1
Gene Aliases: 1110054L24Rik; AU018540; AU019789; HSPC216; NIPA; RGD1561099; ZC3HC1
UniProt ID: (Human) Q86WB0, (Mouse) Q80YV2
Entrez Gene ID: (Human) 51530, (Rat) 296957, (Mouse) 232679
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support