Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BD73; EAR-1; nuclear receptor protein; nuclear receptor Rev-ErbA-alpha; Nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; orphan nuclear receptor; rev-erb alpha; Rev-erba; rev-erbA alpha; rev-erbA(alpha); Rev-erbA-alpha; Rev-ErbA-alpha protein; Rev-ErbAalpha; REV-ERBalpha; Reverbaalpha; V-erbA-related protein 1; V-erbA-related protein EAR-1
Gene Aliases: A530070C09Rik; ear-1; EAR1; HREV; NR1D1; R75201; REV-ERBAALPHA; THRA1; THRAL
UniProt ID: (Human) P20393, (Rat) Q63503, (Mouse) Q3UV55
Entrez Gene ID: (Human) 9572, (Rat) 252917, (Mouse) 217166
Molecular Function:
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support