Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c15; clone 15; H M NPD011; MNUDC protein; nuclear distribution C homolog; nuclear distribution gene C homolog; Nuclear distribution protein C homolog; Nuclear migration protein nudC; nudC nuclear distribution protein; OTTHUMP00000004405; SIG-92; Silica-induced gene 92 protein
Gene Aliases: c15; HNUDC; MNUDC; NPD011; NUDC; SIG-92; Silg92
UniProt ID: (Human) Q9Y266, (Mouse) O35685, (Rat) Q63525
Entrez Gene ID: (Human) 10726, (Mouse) 18221, (Rat) 29648
Molecular Function: microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support