Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
FIGURE: 1 / 4
PDGFRA is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha); Alpha-type platelet-derived growth factor receptor; Beta platelet-derived growth factor receptor; Beta-type platelet-derived growth factor receptor; CD140 antigen-like family member A; CD140 antigen-like family member B; CD140a; CD140a antigen; CD140b; MGC74795; PDGF alpha chain; PDGF beta chain; PDGF Receptor alpha; PDGF-R-alpha; PDGF-R-beta; PDGFR-1; PDGFR-2; pdgfr-a; PDGFR-alpha; PDGFR-beta; PDGFRA/BCR fusion; PGFRB; Platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor 2; Platelet-derived growth factor receptor alpha; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; platelet-derived growth factor receptor, alpha polypeptide; platelet-derived growth factor receptor, beta polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein
Gene Aliases: AI115593; AI528809; CD140A; CD140B; GAS9; IBGC4; IMF1; JTK12; KOGS; PDGFR; PDGFR-1; PDGFR-2; PDGFR1; PDGFR2; PDGFRA; PDGFRB; PENTT; RHEPDGFRA
UniProt ID: (Human) P16234, (Human) P09619, (Mouse) P26618, (Mouse) P05622
Entrez Gene ID: (Human) 5156, (Human) 5159, (Mouse) 18595, (Mouse) 18596
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support