Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of PLD1 only when phosphorylated at Thr147.
Phosphatidylcholine (PC)-specific phospholipases D (PLDs; EC 3.1.4.4) catalyze the hydrolysis of PC to produce phosphatidic acid and choline. A range of agonists acting through G protein-coupled receptors and receptor tyrosine kinases stimulate this hydrolysis. PC-specific PLD activity has been implicated in numerous cellular pathways, including signal transduction, membrane trafficking, and the regulation of mitosis (Hammond et al., 1995).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Choline phosphatase 1; hPLD1; mPLD1; phophatidylcholine-specific; Phoshpolipase D gene 1 probably with two splicing variants A and B; Phosphatidylcholine-hydrolyzing phospholipase D1; phospholipase D gene 1, probably with two splicing variants A and B; Phospholipase D1; phospholipase D1, phosphatidylcholine-specific; PLD 1; rPLD1
Gene Aliases: AA536939; C85393; PLD1; Pld1a; Pld1b; Plda; Pldb
UniProt ID: (Human) Q13393, (Rat) P70496
Entrez Gene ID: (Human) 5337, (Rat) 25096, (Mouse) 18805
Molecular Function:
phospholipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support