Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
This antibody is predicted to react with mouse and rat based on sequence homology.
Protein phosphatase-1 (PP1) is one of the main eukaryotic serine/threonine phosphatases. The protein encoded by this gene binds to the catalytic subunit of PP1, strongly inhibiting its activity. Ten related pseudogenes have been found throughout the human genome. Several splice variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IPP-2; phosphatase inhibitor 2; phosphoprotein phosphatase; protein phosphatase 1, regulatory (inhibitor) subunit 2; protein phosphatase 1, regulatory subunit 2; Protein phosphatase inhibitor 2
Gene Aliases: 0610025N14Rik; 2310007G06Rik; 4930440J04Rik; 5430408E15Rik; D16Ertd248e; IPP-2; IPP2; PPP1R2
UniProt ID: (Human) P41236, (Mouse) Q9DCL8, (Rat) P50411
Entrez Gene ID: (Human) 5504, (Mouse) 66849, (Rat) 192361
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support