Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The antibody detects a human recombinant Prion protein after phosphorylation by Cdk5/p25 complex. In addition, the antibody may detect aggregrated forms of prion in human PC3 cells treated with Calyculin A.
Human prion protein (PrP), also known as PRNP, is a ubiquitously expressed GPI-anchored cell surface glycoprotein associating with lipid raft components and functioning as a signaling molecule. PrP plays a role in apoptosis in a cell context-dependent manner, is involved in proliferation of epithelial cells and in distribution of junction-associated proteins in human enterocytes. Conversion of this normal cellular prion protein (PrPc) into an abnormal conformer (PrPSc) is the crucial step associated with triggering the pathogenesis of the prion neurodegenerative disorders, such as the Creutzfeld-Jakob disease (CJD). Whereas PrPc is rich in alpha-helices, the PrPSc form has higher content of beta-sheets and is resistant to proteinase K.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alternative prion protein; AltPrP; ASCR; BSE; CD230; CD230 antigen; Major prion protein; MGC26679; prion protein, PrP; prion protein, structural; prion-related protein; PrP; PrP27-30; PrP33-35C; TSE
Gene Aliases:
AA960666; AI325101; ALTPRP; ASCR; CD230; CJD; GSS; KURU; p27-30; PRIP; Prn; Prn-i; Prn-p; PRNP; PRP; PrP27-30; PrP33-35C; PrP
UniProt ID: (Human) F7VJQ1, (Mouse) P04925, (Rat) P13852
Entrez Gene ID: (Human) 5621, (Mouse) 19122, (Rat) 24686
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support