Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
Keep as concentrated solution.
Positive Control: 293T (100 uM etoposide treatment for 2 hrs).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Rad17 is essential for sustained cell growth, maintenance of chromosomal stability, and ATR-dependent checkpoint activation upon DNA damage. It has a weak ATPase activity required for binding to chromatin. Rad17 also participates in the recruitment of the RAD1-RAD9-HUS1 complex and RHNO1 onto chromatin, and in CHEK1 activation. May also serve as a sensor of DNA replication progression, and may be involved in homologous recombination.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell cycle checkpoint protein (RAD17); Cell cycle checkpoint protein RAD17; FLJ41520; hRad17; RAD1 homolog; RAD17 (Ser647); RAD17 homolog; Rad17 like protein; Rad17-like protein; RF-C activator 1 homolog; RF-C/activator 1 homolog
Gene Aliases: CCYC; HRAD17; R24L; RAD17; RAD17SP; RAD24
UniProt ID: (Human) O75943
Entrez Gene ID: (Human) 5884
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support