Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with Monkey, Rat and Mouse
Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called “Recombinant polyclonal antibody”, this product is now rebranded as “Recombinant Superclonal™ antibody”. The physical product and the performance remain unchanged.
The p90 ribosomal S6 kinases (RSKs) comprise a family of serine/threonine kinases that lie at the terminus of the ERK pathway. In humans, the RSK family consists of four isoforms (RSK1 to-4). RSK family members are unusual among serine/threonine kinases in that they contain two distinct kinase domains, both of which are catalytically functional. Theses kinase domains are activated in a sequential manner by a series of phosphorylations. RSK regulates gene expression via association and phosphorylation of transcriptional regulators including c-Fos, estrogen receptor, NFkappaB/IkappaB alpha, cAMP-response element-binding protein (CREB). ERK activates the C-terminal kinase of RSK, leading to activation of the N-terminal kinase. Members of the RSK family are present in the cytoplasm as well as the nucleus. Addition of growth factor to the cells results in the activation of both cytosolic and nuclear RSK and the translocation of the cytosolic RSK into the nucleus upon activation. The activation and nuclear translocation of RSK result in phosphorylation and activation of transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 90 kDa ribosomal protein S6 kinase 1; 90 kDa ribosomal protein S6 kinase 2; 90 kDa ribosomal protein S6 kinase 3; 90 kDa ribosomal protein S6 kinase 4; 90 kDa ribosomal protein S6 kinase 6; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); Insulin-stimulated protein kinase 1; ISPK-1; MAP kinase-activated protein kinase 1a; MAP kinase-activated protein kinase 1b; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1a; MAPK-activated protein kinase 1b; MAPK-activated protein kinase 1c; MAPKAP kinase 1a; MAPKAP kinase 1b; MAPKAP kinase 1C; MAPKAPK-1a; MAPKAPK-1b; MAPKAPK1D; mitogen- and stress-activated protein kinase 2; mitogen-activated protein kinase-activated protein kinase 1C; Nuclear mitogen- and stress-activated protein kinase 2; p90 rsk; p90-RSK 1; p90-RSK 2; p90-RSK 3; p90-RSK 6; p90RSK1; p90RSK6; p90S6K; pp90RSK2; pp90RSK3; pp90RSK4; Ribosomal protein kinase B; ribosomal protein S6 kinase alpha 1; ribosomal protein S6 kinase alpha 4; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase alpha-2; Ribosomal protein S6 kinase alpha-3; Ribosomal protein S6 kinase alpha-4; Ribosomal protein S6 kinase alpha-6; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 3; ribosomal protein S6 kinase, 90kDa, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 6; Ribosomal S6 kinase 1; Ribosomal S6 kinase 2; Ribosomal S6 kinase 3; Ribosomal S6 kinase 4; RP11-393H10.3; RSK-1; RSK-2; RSK-3; RSK-4; RSKB; S6K-alpha 1; S6K-alpha 6; S6K-alpha-1; S6K-alpha-2; S6K-alpha-3; S6K-alpha-4; S6K-alpha-6
Gene Aliases: CLS; HU-1; HU-2; HU-3; ISPK-1; ISPK1; MAPKAPK1A; MAPKAPK1B; MAPKAPK1C; MRX19; MSK2; p90-RSK2; p90-RSK3; p90Rsk; p90RSK2; pp90RSK2; pp90RSK3; PP90RSK4; RPS6KA1; RPS6KA2; RPS6KA3; RPS6KA4; RPS6KA6; RSK; RSK-B; RSK1; RSK2; RSK3; RSK4; S6K-alpha; S6K-alpha-4; S6K-alpha2; S6K-alpha3
UniProt ID: (Human) Q15418, (Human) Q15349, (Human) P51812, (Human) O75676, (Human) Q9UK32
Entrez Gene ID: (Human) 6195, (Human) 6196, (Human) 6197, (Human) 8986, (Human) 27330
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support