Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Antibody detects endogenous levels of SAR1A only when phosphorylated at Thr139.
Involved in transport from the endoplasmic reticulum to the Golgi apparatus. Required to maintain SEC16A localization at discrete locations on the ER membrane perhaps by preventing its dissociation. SAR1A-GTP-dependent assembly of SEC16A on the ER membrane forms an organized scaffold defining endoplasmic reticulum exit sites (ERES). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COPII-associated small GTPase; GTP-binding protein SAR1a; SAR1 gene homolog A; SAR1 homolog A; SAR1a gene homolog 1; secretion associated, Ras related GTPase 1A; Secretion-associated Ras-related GTPase 1A; Small COPII coat GTPase SAR1A
Gene Aliases: 1600019H17Rik; masra2; SAR1; SAR1A; SARA; SARA1
UniProt ID: (Human) Q9NR31
Entrez Gene ID: (Human) 56681, (Mouse) 20224, (Rat) 361842
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support