Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The antigen is homologous with the corresponding sequence in mouse.
Specificity of this antibody: SCNN1B.
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception. Heterotetramer of two alpha, one beta and one gamma subunit. A delta subunit can replace the alpha subunit. Interacts with the WW domains of NEDD4, NEDD4L, WWP1 and WWP2. Defects in SCNN1B are a cause of autosomal recessive pseudohypoaldosteronism type 1 (PHA1) [MIM:264350]. PHA1 is a rare salt wasting disease resulting from target organ unresponsiveness to mineralocorticoids. There are 2 forms of PHA1: the autosomal recessive form that is severe, and the dominant form which is more milder and due to defects in mineralocorticoid receptor. Autosomal recessive PHA1 is characterized by an often fulminant presentation in the neonatal period with dehydration, hyponatraemia, hyperkalaemia, metabolic acidosis, failure to thrive and weight loss. Defects in SCNN1B are a cause of Liddle syndrome. It is an autosomal dominant disorder characterized by pseudoaldosteronism and hypertension associated with hypokalemic alkalosis. The disease is caused by constitutive activation of the renal epithelial sodium channel.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amiloride-sensitive sodium channel subunit beta; Beta ENaC; Beta-ENaC; Beta-NaCH; ENaC beta; Epithelial Na(+) channel subunit beta; Nonvoltage-gated sodium channel 1 subunit beta; SCNEB; SCNN 1B; sodium channel, nonvoltage-gated 1, beta; sodium channel, nonvoltage-gated, type I, beta
Gene Aliases: RNENACB; Scnn1b
UniProt ID: (Mouse) Q9WU38, (Rat) P37090
Entrez Gene ID: (Mouse) 20277, (Rat) 24767
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support