Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of Semaphorin 4B only when phosphorylated at Ser809.
Inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sema C; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4B; Semaphorin-4B; Semaphorin-C
Gene Aliases: KIAA1745; mKIAA1745; SEMA4B; SEMAC; SemC; UNQ749/PRO1480
UniProt ID: (Human) Q9NPR2, (Mouse) Q62179
Entrez Gene ID: (Human) 10509, (Mouse) 20352
Molecular Function: membrane-bound signaling molecule intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support