Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phospho-SHC3 (Tyr424) Polyclonal Antibody detects endogenous levels of SHC3 only when phosphorylated at Tyr424.
Signaling adapter that couples activated growth factor receptors to signaling pathway in neurons. Involved in the signal transduction pathways of neurotrophin-activated Trk receptors in cortical neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-Shc; Neuronal Shc; Protein Rai; SH2 domain protein C3; SHC (Src homology 2 domain containing) transforming protein 3; SHC-transforming protein 3; SHC-transforming protein C; Shc3 p51; src homology 2 domain-containing transforming protein C3; Src homology 2 domain-containing-transforming protein C3
Gene Aliases: N-Shc; NSHC; RAI; SHC3; SHCC
UniProt ID: (Human) Q92529, (Mouse) Q3ZAX2
Entrez Gene ID: (Human) 53358, (Rat) 114858, (Mouse) 20418
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support