Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Phospho-SMAD1/SMAD5/SMAD9 (Ser463, Ser465) Polyclonal Antibody detects endogenous levels of SMAD1/SMAD5/SMAD9 only when phosphorylated at Ser463, Ser465.
The protein encoded by SMAD1/SMAD5/SMAD8/SMAD9 belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein mediates the signals of the bone morphogenetic proteins (BMPs), which are involved in a range of biological activities including cell growth, apoptosis, morphogenesis, development and immune responses. In response to BMP ligands, this protein can be phosphorylated and activated by the BMP receptor kinase. The phosphorylated form of this protein forms a complex with SMAD4, which is important for its function in the transcription regulation. This protein is a target for SMAD-specific E3 ubiquitin ligases, such as SMURF1 and SMURF2, and undergoes ubiquitination and proteasome-mediated degradation. Alternatively spliced transcript variants encoding the same protein have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BSP-1; JV4-1; JV5-1; MAD (mothers against decapentaplegic, Drosophila) homolog 1; MAD homolog 1; MAD homolog 5; MAD homolog 9; MAD homolog1 (mothers against decapentaplegic, Drosophila); MAD, mothers against decapentaplegic homolog 1; MAD, mothers against decapentaplegic homolog 5; Mad-related protein 1; Madh6; Mothers against decapentaplegic homolog 1; Mothers against decapentaplegic homolog 5; Mothers against decapentaplegic homolog 9; mothers against decapentaplegic, drosophila, homolog of, 5; Mothers against decapentaplegic, drosophila, homolog of, 9; mothers against DPP homolog 1; mothers against DPP homolog 5; mothers against DPP homolog 9; SMA- and MAD-related protein 5; SMAD; SMAD 1; SMAD 5; SMAD 9; SMAD family member 1; SMAD family member 5; SMAD family member 9; SMAD, mothers against DPP homolog 1; SMAD, mothers against DPP homolog 5; SMAD, mothers against DPP homolog 9; Smad8; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1
Gene Aliases: BSP-1; BSP1; DWFC; JV4-1; JV41; JV5-1; Mad1; MADH1; MADH5; MADH6; Madh8; MADH9; MADR1; PPH2; SMAD1; SMAD5; SMAD8; SMAD8/9; SMAD8A; SMAD8B; SMAD9
UniProt ID: (Human) Q15797, (Human) Q99717, (Human) O15198, (Rat) P97588, (Rat) Q9R1V3, (Rat) O54835
Entrez Gene ID: (Human) 4086, (Human) 4090, (Human) 4093, (Rat) 25671, (Rat) 59328, (Rat) 85435
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support