Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phospho-SRC1 (Thr1179) Polyclonal Antibody detects endogenous levels of SRC1 only when phosphorylated at Thr1179.
Clone 1135 recognizes SRC-1, steroid-receptor coactivator-1. SRC-1 is a member of a gene family that functions in the full transcriptional activation of the steroid hormone receptor superfamily and contains intrinsic histone acetyltransferase activity. SRC-1 has a molecular weight of 160 kDa and has a basic helix-loop-helix (bHLH)-PAS domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASV; bHLHe74; c-SRC; Class E basic helix-loop-helix protein 74; Hin-2 protein; mNRC-1; NCoA-1; Nuclear receptor coactivator 1; Nuclear receptor coactivator protein 1; p60-Src; PAX3/NCOA1 fusion protein; Protein Hin-2; Renal carcinoma antigen NY-REN-52; RIP160; SRC-1; SRC-1 (-Q); SRC-a/NCoA-1; Steroid receptor coactivator 1; steroid receptor coactivator-1
Gene Aliases: bHLHe42; BHLHE74; F-SRC-1; KAT13A; NCOA1; RIP160; SRC-1; SRC1
UniProt ID: (Human) Q15788, (Mouse) P70365
Entrez Gene ID: (Human) 8648, (Mouse) 17977
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support