Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phospho-STMN2 (Ser73) Polyclonal Antibody detects endogenous levels of STMN2 only when phosphorylated at Ser73.
STMN2, also known as SCG-10, is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin. STMN2 may play a role in neuronal differentiation, and in modulating membrane interaction with the cytoskeleton during neurite outgrowth. Reductions in the expression of this gene have been associated with Down's syndrome and Alzheimer's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neuron-specific growth-associated protein; neuronal growth-associated protein (silencer element); Protein SCG10; SGC10; Stathmin-2; stathmin-like 2; superior cervical ganglia, neural specific 10; Superior cervical ganglion-10 protein; superiorcervical ganglia, neural specific 10
Gene Aliases: AI159727; SCG10; SCGN10; Stmb2; STMN2
UniProt ID: (Human) Q93045, (Rat) P21818, (Mouse) P55821
Entrez Gene ID: (Human) 11075, (Rat) 84510, (Mouse) 20257
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support