Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of Syndecan 4 only when phosphorylated at Ser179.
Syndecans are type I integral membrane proteoglycans that are involved in cell-extracellular matrix adhesion and growth factor binding. Syndecan 1 is a matrix receptor which binds to Collagens, Fibronectin and Thrombospondin. Syndecan 1 and Syndecan 3 interact with MK (midkine), a growth/differentiation factor involved in embryogenesis. Syndecan 2 is highly expressed at areas of high morphogenetic activity, such as epithelial-mesenchymal interfaces and the prechondrogenic and preosteogenic mesenchymal condensations. Syndecan 4 functions cooperatively with integrins in the processes of cell spreading and focal adhesion assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amphiglycan; MGC22217; ryudocan amphiglycan; Ryudocan core protein; Ryudocan/syndecan 4; SYND4; syndecan 4 (amphiglycan, ryudocan); syndecan proteoglycan 4; Syndecan-4
Gene Aliases: AA959608; AW108331; RATRYUDOCA; RYUDOCA; ryudocan; S4; SDC4; SYND4; syndecan-4
UniProt ID: (Human) P31431, (Rat) P34901, (Mouse) O35988
Entrez Gene ID: (Human) 6385, (Rat) 24771, (Mouse) 20971
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support