Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • TAK1 Antibodies

            Invitrogen

            Phospho-TAK1 (Thr184, Thr187) Polyclonal Antibody

            View all (52) TAK1 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite Phospho-TAK1 (Thr184, Thr187) Polyclonal Antibody

            • Antibody Testing Data (2)
            Application
            Phospho-TAK1 (Thr184, Thr187) Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            Phospho-TAK1 (Thr184, Thr187) Antibody (PA5-105123) in WB

            Western blot was performed using Anti-Phospho-TAK1 (Thr184, Thr187) Polyclonal Antibody (Product # PA5-105123) and a ~65 kDa band corresponding to Mitogen-activated protein kinase kinase kinase 7 was observed along with two uncharacterised band (*) at ~260 kDa and ~40 kDa in the cell lines tested. Fresh Whole cell extracts (40 µg lysate) of HEK-293 (Lane 1) and IL1a treated HEK-293 (Lane 2) were electrophor... View More
            View Product

            Phospho-TAK1 (Thr184, Thr187) Antibody in Western Blot (WB)
            Phospho-TAK1 (Thr184, Thr187) Antibody in Western Blot (WB)

            Product Details

            PA5-105123

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2,000
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            A synthesized peptide derived from human MAP3K7(Accession O43318), corresponding to amino acid residues around phosphorylated Thr187.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            sequential chromatography

            Storage buffer

            PBS, pH 7.4, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2816596

            Product Specific Information

            Antibody detects endogenous levels of TAK1 only when phosphorylated at Thr184/187.

            Target Information

            TAK1 (also known as MAP3K7), a part of the serine/threonine kinase family, is a versatile protein with many signaling functions. Originally identified as a TGF- beta-activated kinase, TAK1 interacts with TGF- beta-receptor and TRAF6 to modulate TGF- beta activation of JNK and p38. It has additional roles in activation of p38 and JNK through Wnt, BMP, and activin signaling pathways as well in response to thyroid hormone, osmotic stress, endothelin, and ephrine. Through activation of von Hippel-Lindau tumor suppressor expression, TAK1 represses PDGF-B, integrin beta1 and integrin beta5, promoting proper wound healing. TAK1 also plays an essential role in IKK activation in numerous signaling pathways including IL-1, IL-6, IL-18, TNF, CD40, TLR, and RIG-I. Activation of the TAK1-IKK pathway requires TAB2/3 and ubiquitination. In this process, TAB2/3 binds to the C-terminal region of TAK1 and becomes polyubiquitinated, TAK1 is autophosphorylated at Thr178, Thr184, Thr187 and Ser192, and finally TAK1 phosphorylates IKK- beta. Additionally, TAK1 represses human telomerase reverse transcriptase suggesting a role in regulation of cell lifespan.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: M3K7; mitogen activated protein kinase kinase kinase 7; Mitogen-activated protein kinase kinase kinase 7; RP1-154G14.1; TGF-beta activated kinase 1; TGF-beta-activated kinase 1; transforming growth factor beta-activated kinase 1; Transforming growth factor-beta-activated kinase 1

            View more View less

            Gene Aliases: B430101B05; C87327; MAP3K7; MEKK7; TAK1; TGF1a

            View more View less

            UniProt ID: (Human) O43318, (Mouse) Q62073, (Rat) P0C8E4

            View more View less

            Entrez Gene ID: (Human) 6885, (Mouse) 26409, (Rat) 313121

            View more View less

            Function(s)

            magnesium ion binding protein kinase activity protein serine/threonine kinase activity MAP kinase kinase kinase activity protein binding ATP binding scaffold protein binding nucleotide binding MAP kinase kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups metal ion binding non-receptor serine/threonine protein kinase protein modifying enzyme

            Process(es)

            activation of MAPKK activity activation of MAPK activity stimulatory C-type lectin receptor signaling pathway positive regulation of T cell cytokine production MyD88-dependent toll-like receptor signaling pathway transcription, DNA-templated apoptotic process transforming growth factor beta receptor signaling pathway Wnt signaling pathway, calcium modulating pathway I-kappaB kinase/NF-kappaB signaling activation of NF-kappaB-inducing kinase activity I-kappaB phosphorylation JNK cascade positive regulation of interleukin-2 production Fc-epsilon receptor signaling pathway positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of JUN kinase activity histone H3 acetylation T cell receptor signaling pathway positive regulation of T cell activation positive regulation of NF-kappaB transcription factor activity stress-activated MAPK cascade nucleotide-binding oligomerization domain containing signaling pathway MAPK cascade angiogenesis neural tube formation regulation of transcription, DNA-templated protein phosphorylation phosphorylation positive regulation of JNK cascade negative regulation of necroptotic process negative regulation of ripoptosome assembly involved in necroptotic process regulation of reactive oxygen species metabolic process negative regulation of reactive oxygen species metabolic process negative regulation of apoptotic signaling pathway
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-v98pj:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0