Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of TCTP only when phosphorylated at Ser46.
TPT1 (translationally controlled tumor protein 1) is a 23 kDa member of the TCTP family of calcium binding proteins. TPT1 is localized in the cytoplasm and widely expressed and serves as a transcriptional activator, calcium transporter, histamine inducer and antiapoptotic caspase 3 inhibitor. TPT1 is a cytokine-like molecule that causes the release of histamine, IL-4 and IL-13 from basophils as well as the secretion of IL-8 and a calcium response in eosinophils. TPT1 plays a pivotal role in allergic diseases and due to its wide distribution in brain, is thought to be involved in neurodegenerative disorders, such as Alzheimer's disease and Down syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 21 kDa polypeptide; 21 kDa polypeptide under translational control; Fortilin; Histamine-releasing factor; HRF; Lens epithelial protein; p21; p23; TCTP; translationally regulated transcript (21 kDa); Translationally-controlled tumor protein
Gene Aliases: HRF; p02; p21; p23; TCTP; TPT1; Trt
UniProt ID: (Human) P13693, (Mouse) P63028, (Rat) P63029
Entrez Gene ID: (Human) 7178, (Mouse) 22070, (Rat) 116646
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support