Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
Antibody detects endogenous levels of TRIM32 only when phosphorylated at Ser339.
TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 72 kDa Tat-interacting protein; bM468K2.2 (tripartite motif protein 32); E3 ubiquitin-protein ligase TRIM32; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; tripartite motif protein 32; tripartite motif-containing 32; Tripartite motif-containing protein 32; zinc finger protein 117; Zinc finger protein HT2A; zinc-finger protein HT2A
Gene Aliases: 1810045E12Rik; 3f3; BBS11; HT2A; LGMD2H; TATIP; TRIM32; Zfp117
UniProt ID: (Human) Q13049, (Mouse) Q8CH72
Entrez Gene ID: (Human) 22954, (Mouse) 69807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support