Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of ZAK only when phosphorylated at Ser165.
ZAK is a leucine zipper-containing mitogen-activated protein related to the MLKs in both sequence and function. Like the MLKs and other MAP3Ks, ZAK activates MAP2Ks. ZAK is a member of the MAPKKK family of signal transduction molecules and encodes a protein with an N-terminal kinase catalytic domain, followed by a leucine zipper motif and a sterile-alpha motif. This magnesium-binding protein forms homodimers and is located in the cytoplasm. The protein mediates gamma radiation signaling leading to cell cycle arrest and activity of this protein plays a role in cell cycle checkpoint regulation in cells. The protein also has pro-apoptotic activity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HCCS-4; Human cervical cancer suppressor gene 4 protein; Leucine zipper- and sterile alpha motif kinase ZAK; Leucine zipper- and sterile alpha motif-containing kinase; Mitogen-activated protein kinase kinase kinase 20; Mitogen-activated protein kinase kinase kinase MLT; Mixed lineage kinase 7; mixed lineage kinase with a leucine zipper and a sterile alpha motif; Mixed lineage kinase-related kinase; mixed lineage kinase-related kinase MRK-beta; MLK-like mitogen-activated protein triple kinase; MLK-related kinase; MRK; sterile alpha motif and leucine zipper containing kinase AZK; Sterile alpha motif- and leucine zipper-containing kinase AZK; sterile-alpha motif and leucine zipper containing kinase AZK
Gene Aliases: AV006891; AZK; B230120H23Rik; HCCS4; MAP3K20; MLK7; mlklak; MLT; MLTK; MLTKalpha; MLTKbeta; MRK; pk; RGD1561394; SFMMP; ZAK
UniProt ID: (Human) Q9NYL2, (Mouse) Q9ESL4
Entrez Gene ID: (Human) 51776, (Mouse) 65964, (Rat) 311743
Molecular Function: immunoglobulin non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support