Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
OPRD1 is an Opioid Receptor that is involved in the control of anxiety, depression, and pain. It is a promising target in the therapy of mood-altering disorders and drug addiction. Delta and kappa opioid receptors function synergistically by forming heterodimers. OPRD1 expression has been reported primarily in the brain. High OPRD1 expression has been documented in the cortex, caudate, hippocampus, nucleus accumbens, and putamen; low OPRD1 expression has been identified in the cerebellum, hypothalamus, substantia nigra, and thalamus. OPRD1 expression is reduced in the putamen and amygdala of Alzheimer's patients and in the putamen in Parkinson's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-OR-1; delta 1 opioid receptor; delta opioid receptor 1; Delta-type opioid receptor; DOR-1; K56; MSL-2; Opioid receptor; Opioid receptor A; opioid receptor, delta 1
Gene Aliases: DOR; DOR-1; mDOR; Nbor; OPRD; OPRD1; Ror-a
UniProt ID: (Human) P41143, (Mouse) P32300, (Rat) P33533
Entrez Gene ID: (Human) 4985, (Mouse) 18386, (Rat) 24613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support